Novus Biologicals
Manufacturer Code:NBP274442AF700
Catalog # N274442A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A5 |
Immunogen | Recombinant protein fragment corresponding to amino acids 782-1121 of human TAF2 (NP_003175) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 150 kDa cofactor of initiator function; 150 kDa cofactor of initiator function CIF150TAFII-150 cofactor of initiator function 150kD subunit TAF(II)150 TAF2 RNA polymerase II TATA box binding protein (TBP)-associated factor TAF2B TAFII150RNA polymerase II TBP-associated factor subunit B TATA box binding protein (TBP)-associated factor RNA polymerase II B 150kD TBP-associated factor 150 kDa Transcription initiation factor TFIID 150 kDa subunit transcription initiation factor TFIID subunit 2; cofactor of initiator function, 150kD subunit; RNA polymerase II TBP-associated factor subunit B; TAF(II)150; TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 150kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD; TATA box binding protein associated factor 2; TBP-associated factor 150 kDa; Transcription initiation factor TFIID 150 kDa subunit; Transcription initiation factor TFIID subunit 2
Gene Aliases: CIF150; MRT40; TAF2; TAF2B; TAFII150
UniProt ID: (Human) Q6P1X5
Entrez Gene ID: (Human) 6873
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.