Invitrogen
Manufacturer Code:PA540710
Catalog # PIPA540710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TAF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 150 kDa cofactor of initiator function; cofactor of initiator function, 150kD subunit; RNA polymerase II TBP-associated factor subunit B; TAF(II)150; TAF2 RNA polymerase II CIF150 TAF2B TAFII150; TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 150kDa; TAF2 RNA polymerase II, TATA box binding protein-associated factor, 150 kD; TATA box binding protein (TBP)-associated factor 150kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD; TATA box binding protein associated factor 2; TBP-associated factor 150 kDa; TBP-associated factor 2; Transcription initiation factor TFIID 150 kDa subunit; Transcription initiation factor TFIID subunit 2; transcription initiation factor TFIID subunit 2-like protein
Gene Aliases: 150kDa; 4732460C16Rik; AI425886; CIF150; fc30f02; fc56a05; MRT40; TAF2; TAF2B; TAFII-150; TAFII150; TAFIIB; wu:fc30f02; wu:fc56a05
UniProt ID: (Human) Q6P1X5, (Zebrafish) Q32PW3
Entrez Gene ID: (Bovine) 781912, (Dog) 475084, (Horse) 100057143, (Rabbit) 100353006, (Human) 6873, (Guinea pig) 100722226, (Rat) 170844, (Mouse) 319944, (Zebrafish) 324486
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.