Abnova
Manufacturer Code:PAB16029
Catalog # 89-118-024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of <i>Saccharomyces cerevisiae</i> TAF2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TAF2.
For Research Use Only
Protein Aliases: TAF150; Taf2p; TAFII-150; TBP-associated factor 150 kDa; TBP-associated factor 2; Transcription initiation factor TFIID subunit 2; TSM-1; TSM1
Gene Aliases: TAF150; TAF2; TSM1; YCR042C; YCR42C; YCR724
UniProt ID: (Baker's yeast) P23255
Entrez Gene ID: (Baker's yeast) 850409
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.