Invitrogen
Manufacturer Code:PA563212
Catalog # PIPA563212
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TAF1D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JOSD3 RAFI41 TAF(I)41 TAFI41 Josephin domain containing 3 RNA polymerase I-specific TBP-associated factor 41 kDa TATA box-binding protein-associated factor 1D TATA box-binding protein-associated factor RNA polymerase I subunit D TBP-associated factor 1D transcription initiation facto; Josephin domain containing 3; RNA polymerase I-specific TBP-associated factor 41 kDa; TAFI41; TATA box binding protein (TBP)-associated factor, RNA polymerase I, D, 41kDa; TATA box binding protein associated factor 1D; TATA box-binding protein-associated factor 1D; TATA box-binding protein-associated factor RNA polymerase I subunit D; TATA-box binding protein associated factor, RNA polymerase I, D; TBP-associated factor 1D; Transcription initiation factor SL1/TIF-IB subunit D
Gene Aliases: JOSD3; RAFI41; TAF(I)41; TAF1D; TAFI41
UniProt ID: (Human) Q9H5J8
Entrez Gene ID: (Human) 79101
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.