Novus Biologicals
Manufacturer Code:NBP274441AF700
Catalog # N274441A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A8 |
Immunogen | Recombinant protein fragment corresponding to amino acids 1-208 of human TAF1B (NP_005671) produced in E.coli. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: RAF1BTATA box binding protein (TBP)-associated factor RNA polymerase I B 63kD RAFI63MGC:9349 RNA polymerase I-specific TBP-associated factor 63 kDa SL1SL1 63kD subunit TAFI63TBP-associated factor 1B TATA box binding protein (TBP)-associated factor RNA polymerase I B 63kDa TATA box-binding protein-associated factor 1B TATA box-binding protein-associated factor RNA polymerase I subunit B Transcription initiation factor SL1/TIF-IB subunit B; RNA polymerase I-specific TBP-associated factor 63 kDa; SL1, 63kD subunit; TAFI63; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa; TATA box binding protein associated factor 1B; TATA box-binding protein-associated factor 1B; TATA box-binding protein-associated factor RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I, B; TBP-associated factor 1B; Transcription initiation factor SL1/TIF-IB subunit B
Gene Aliases: MGC:9349; RAF1B; RAFI63; SL1; TAF1B; TAFI63
UniProt ID: (Human) Q53T94
Entrez Gene ID: (Human) 9014
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.