Invitrogen
Manufacturer Code:MA527453
Catalog # PIMA527453
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7A11 |
Immunogen | Full length human recombit protein of TAF1A produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC:17061 RAFI48 SL1 TAFI48 TATA box-binding protein-associated factor RNA polymerase I subunit A RNA polymerase I-specific TBP-associated factor 48 kDa SL1 48kD subunit TATA box binding protein (TBP)-associated factor RNA polymerase I A 48kD TATA box binding protein (TBP)-associated factor RNA polymerase I A 48kDa TATA box-binding protein-associated factor 1A TATA-box binding protein associated factor RNA polymerase I A TBP-associated factor 1A transcription factor SL1 transcription initiation factor SL1/TIF-IB subunit A; RNA polymerase I-specific TBP-associated factor 48 kDa; SL1, 48kD subunit; TAFI48; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa; TATA box binding protein associated factor 1A; TATA box-binding protein-associated factor 1A; TATA box-binding protein-associated factor RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I, A; TBP-associated factor 1A; Transcription factor SL1; Transcription initiation factor SL1/TIF-IB subunit A
Gene Aliases: MGC:17061; RAFI48; SL1; TAF1A; TAFI48
UniProt ID: (Human) Q15573
Entrez Gene ID: (Human) 9015
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.