Invitrogen
Manufacturer Code:PA528429
Catalog # PIPA528429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1787 and 1849 of Human TAF172 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase BTAF1; B-TFIID transcription factor-associated 170 kDa subunit; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1 homolog, S. cerevisiae); Mot1 homolog; MOT1 TAF(II)170 TAF172 TAFII170 ATP-dependent helicase BTAF1 B-TFIID transcription factor-associated 170 kDa subunit TATA-binding protein-associated factor 172 TBP-associated factor 172; TAF(II)170; TAF-172; TATA-binding protein-associated factor 172; TBP-associated factor 172
Gene Aliases: BTAF1; MOT1; TAF(II)170; TAF172; TAFII170
UniProt ID: (Human) O14981
Entrez Gene ID: (Human) 9044
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.