Novus Biologicals
Manufacturer Code:NBP288402
Catalog # NB036024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide corresponding to a region of Mouse TAF12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20kDa TAF12 RNA polymerase II TATA box binding protein (TBP)-associated factor TAF15 TAF2J TAFII-20/TAFII-15 TAFII20TAFII20/TAFII15 TATA box binding protein (TBP)-associated factor RNA polymerase II J 20kD Transcription initiation factor TFIID 20/15 kDa subunits transcription initiation factor TFIID subunit 12; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa; TAFII-20/TAFII-15; TAFII20/TAFII15; TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD; TATA box binding protein associated factor 12; Transcription initiation factor TFIID 20/15 kDa subunits; Transcription initiation factor TFIID subunit 12
Gene Aliases: TAF12; TAF15; TAF2J; TAFII20
UniProt ID: (Human) Q16514
Entrez Gene ID: (Human) 6883
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.