Proteintech
Manufacturer Code:123531AP150UL
Catalog # 50-172-7917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TAF12 Fusion Protein Ag3017 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kDa; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa; TAF12 TAF15 TAF2J TAFII 20/TAFII 15 TAFII20 TAFII20/TAFII15; TAFII-20; TAFII-20/TAFII-15; TAFII20; TAFII20/TAFII15; TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD; TATA box binding protein associated factor 12; Transcription initiation factor TFIID 20 kDa subunits; Transcription initiation factor TFIID 20/15 kDa subunits; Transcription initiation factor TFIID subunit 12
Gene Aliases: 20kDa; 2810422D08Rik; AW557038; TAF12; TAF15; TAF2J; TAFII20
UniProt ID: (Human) Q16514, (Mouse) Q8VE65
Entrez Gene ID: (Human) 6883, (Mouse) 66464
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.