Invitrogen
Manufacturer Code:PA540382
Catalog # PIPA540382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TADA3L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA3 homolog; ADA3-like protein; alteration/deficiency in activation 3; hADA3; mADA3; STAF54; Transcriptional adapter 3; Transcriptional adapter 3-like; Transcriptional adaptor 3 ADA3 FLJ20221 FLJ21329 hADA3 NGG1 TADA3L
Gene Aliases: 1110004B19Rik; ADA3; AI987856; fa03f04; hADA3; NGG1; STAF54; TADA3; TADA3L; wu:fa03f04; zgc:63596
UniProt ID: (Human) O75528, (Bovine) Q5EAE2, (Rat) Q4V8F5, (Mouse) Q8R0L9, (Zebrafish) Q7SY21
Entrez Gene ID: (Dog) 476541, (Human) 10474, (Rabbit) 100347359, (Horse) 100052239, (Bovine) 510184, (Guinea pig) 100719194, (Rat) 362414, (Mouse) 101206, (Zebrafish) 334785
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.