Invitrogen
Manufacturer Code:PA527591
Catalog # PIPA527591
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 251 and 432 of Human TADA3L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA3 homolog; ADA3 NGG1 TADA3L hADA3 ADA3 homolog ADA3-like protein STAF54 transcriptional adapter 3; ADA3-like protein; alteration/deficiency in activation 3; hADA3; STAF54; Transcriptional adapter 3; Transcriptional adapter 3-like
Gene Aliases: ADA3; hADA3; NGG1; STAF54; TADA3; TADA3L
UniProt ID: (Human) O75528
Entrez Gene ID: (Human) 10474
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.