Novus Biologicals
Manufacturer Code:NBP190243
Catalog # NBP190243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LAKEEVSRQELRQRVRMADNEVMDAFRKIMAARQKKRTPTKKEKDQAWKTLKERESILKLLDG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA3 ADA3 homolog alteration/deficiency in activation 3 epididymis secretory sperm binding protein FLJ20221 FLJ21329 hADA3 NGG1 STAF54 TADA3L transcriptional adapter 3 Transcriptional adapter 3-like transcriptional adaptor 3; ADA3 homolog; ADA3-like protein; alteration/deficiency in activation 3; hADA3; STAF54; Transcriptional adapter 3; Transcriptional adapter 3-like
Gene Aliases: ADA3; hADA3; NGG1; STAF54; TADA3; TADA3L
UniProt ID: (Human) O75528
Entrez Gene ID: (Human) 10474
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.