Abnova
Manufacturer Code:H00010474K
Catalog # 89-201-656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TADA3L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TADA3L peptide using ARM Technology.
For Research Use Only
Protein Aliases: ADA3 FLJ20221 FLJ21329 hADA3; ADA3 homolog; ADA3-like protein; alteration/deficiency in activation 3; hADA3; STAF54; Transcriptional adapter 3; Transcriptional adapter 3-like
Gene Aliases: ADA3; hADA3; NGG1; STAF54; TADA3; TADA3L
UniProt ID: (Human) O75528
Entrez Gene ID: (Human) 10474
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.