Invitrogen
Manufacturer Code:PA537069
Catalog # PIPA537069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 248-293 of human TADA2L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA2-like protein; Transcriptional adapter 2-alpha; Transcriptional adapter 2-alpha Transcriptional adapter 2-like ADA2-like protein TADA2A TADA2L KL04P; Transcriptional adapter 2-like; transcriptional adaptor 2 alpha
Gene Aliases: ADA2; ADA2A; AV319371; D030022J10Rik; hADA2; KL04P; TADA2A; TADA2L
UniProt ID: (Human) O75478, (Rat) Q6AYE3, (Mouse) Q8CHV6
Entrez Gene ID: (Human) 6871, (Rat) 360581, (Mouse) 217031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.