Invitrogen
Manufacturer Code:PA5113572
Catalog # PIPA5113572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal region of human TADA2L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA2 ADA2A ADA2-like protein AV319371 D030022J10Rik hADA2 KL04P Tada2a Tada2l Transcriptional adapter 2-alpha Transcriptional adapter 2-like transcriptional adaptor 2 alpha transcriptional adaptor 2A; ADA2-like protein; Transcriptional adapter 2-alpha; Transcriptional adapter 2-like; transcriptional adaptor 2 alpha
Gene Aliases: ADA2; ADA2A; hADA2; KL04P; TADA2A; TADA2L
UniProt ID: (Human) O75478
Entrez Gene ID: (Human) 6871
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.