Proteintech
Manufacturer Code:112991AP150UL
Catalog # 50-172-7040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TADA2L Fusion Protein Ag1817 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA2 ADA2 like protein ADA2A hADA2 KL04P TADA2A TADA2L Transcriptional adapter 2 like; ADA2-like protein; Transcriptional adapter 2-alpha; Transcriptional adapter 2-like; transcriptional adaptor 2 alpha
Gene Aliases: ADA2; ADA2A; AV319371; D030022J10Rik; hADA2; KL04P; TADA2A; TADA2L
UniProt ID: (Human) O75478, (Rat) Q6AYE3, (Mouse) Q8CHV6
Entrez Gene ID: (Human) 6871, (Rat) 360581, (Mouse) 217031
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.