Invitrogen
Manufacturer Code:PA569843
Catalog # PIPA569843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human TADA1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SPT3-associated factor 42; SPT3-associated factor 42 (STAF42); STAF42; TADA1 TADA1L; Transcriptional adapter 1; Transcriptional adapter 1-like protein; transcriptional adaptor 1 (HFI1 homolog, yeast); transcriptional adaptor 1 (HFI1 homolog, yeast) like; transcriptional adaptor 1- like
Gene Aliases: 2900026B15Rik; ADA1; D1Ertd251e; hADA1; HFI1; RGD1306774; STAF42; TADA1; TADA1L; zgc:85851
UniProt ID: (Human) Q96BN2, (Bovine) A6QR06, (Rat) Q5BJQ7, (Mouse) Q99LM9, (Zebrafish) Q6NWA8
Entrez Gene ID: (Rabbit) 100358442, (Dog) 480090, (Pig) 100155439, (Horse) 100056344, (Human) 117143, (Bovine) 521918, (Guinea pig) 100734529, (Rat) 360874, (Mouse) 27878, (Zebrafish) 402899
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.