Proteintech
Manufacturer Code:203371AP150UL
Catalog # 50-173-2835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TADA1L Fusion Protein Ag14118 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0764 RP1 9E21.4 SPT3 associated factor 42 STAF42 TADA1 TADA1L Transcriptional adapter 1; SPT3-associated factor 42; SPT3-associated factor 42 (STAF42); STAF42; Transcriptional adapter 1; Transcriptional adapter 1-like protein; transcriptional adaptor 1 (HFI1 homolog, yeast); transcriptional adaptor 1 (HFI1 homolog, yeast) like
Gene Aliases: 2900026B15Rik; ADA1; D1Ertd251e; hADA1; HFI1; STAF42; TADA1; TADA1L
UniProt ID: (Human) Q96BN2, (Mouse) Q99LM9
Entrez Gene ID: (Human) 117143, (Mouse) 27878
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.