Invitrogen
Manufacturer Code:PA534248
Catalog # PIPA534248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from N-terminal extracellular domain of human TACR3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neurokinin B receptor; neurokinin beta receptor; Neuromedin-K receptor; NK-3 receptor; NK-3R; NKR; Tachykinin receptor 3; TACR3 HH11 Nmk-r NK3R NKR Tachykinin receptor 3 Neurokinin B Neurokinin B receptor NK-3 receptor NK-3R TAC3R Neurokinin 3 receptor Neurokinin beta receptor Neuromedin-K receptor TAC3RL
Gene Aliases: HH11; NK-3R; NK3R; NKR; TAC3R; TAC3RL; TACR3
UniProt ID: (Human) P29371
Entrez Gene ID: (Human) 6870
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.