Invitrogen
Manufacturer Code:PA520392
Catalog # PIPA520392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide from near the center of human NK3R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neurokinin 3 receptor; Neurokinin B receptor; neurokinin beta receptor; neuromedin K receptor; Neuromedin K receptor (Neurokinin B/Tachikin 3); Neuromedin-K receptor; NK-3 receptor; NK-3R; NK-3R NK3R NKR TAC3RL NK-3 receptor neurokinin B receptor neurokinin beta receptor neuromedin-K receptor; Nk3 receptor; NKR; tachikin receptor 3; Tachykinin receptor 3
Gene Aliases: HH11; NK-3R; NK3R; NKR; Nmkr; TAC3R; TAC3RL; TACR3
UniProt ID: (Human) P29371, (Mouse) P47937, (Rat) P16177
Entrez Gene ID: (Human) 6870, (Mouse) 21338, (Rat) 24808
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.