Invitrogen
Manufacturer Code:PA118309
Catalog # 01-670-562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the C-terminal residues A(434) S T T S S F I S S P Y T S V D E Y S(452) of rat NK-3 receptor protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neurokinin B receptor; Neuromedin K receptor (Neurokinin B/Tachikin 3); Neuromedin K receptor NKR Neurokinin B receptor NK-3 receptor NK-3R Tachykinin receptor 3 Tacr3 Tac3r; Neuromedin-K receptor; NK-3 receptor; NK-3R; NKR; tachikin receptor 3; Tachykinin receptor 3
Gene Aliases: Nkr; Nmkr; Tac3r; Tacr3
UniProt ID: (Rat) P16177
Entrez Gene ID: (Rat) 24808
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.