Invitrogen
Manufacturer Code:PA533967
Catalog # PIPA533967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from C-terminus of human Neurokinin A Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neurokinin 2 receptor; Neurokinin A receptor; NK-2 receptor; NK-2R; seven transmembrane helix receptor; SKR; substance K receptor; Substance-K receptor; Tachykinin receptor 2; TACR2 Neurokinin 2 receptor NK-2 receptor Neurokinin A receptor NK2 NK2R Tachykinin receptor 2 Substance-K receptor NK-2R SK receptor SKR TAC2R NKNAR Substance K receptor
Gene Aliases: NK2R; NKNAR; SKR; TAC2R; TACR2
UniProt ID: (Human) P21452
Entrez Gene ID: (Human) 6865
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.