Invitrogen
Manufacturer Code:PA116713
Catalog # APPA6713
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Raised against a synthetic peptide at the c-terminus of rat NK-1 conjugated to bovine thyroglobulin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neurokinin 1 receptor; neurokinin receptor 1; neurokinin-1 receptor; NK-1 receptor; NK-1R; Nk1 receptor; NK1 receptor NK1R SPR Substance P receptor TAC1R Tachykinin receptor 1 TACR1; SPR; substance P receptor; Substance-P receptor; Tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor)
Gene Aliases: NK1R; NKIR; SPR; TAC1R; TACR1
UniProt ID: (Human) P25103, (Guinea pig) P30547, (Mouse) P30548, (Rat) P14600
Entrez Gene ID: (Human) 6869, (Guinea pig) 100135626, (Mouse) 21336, (Rat) 24807
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.