Novus Biologicals
Manufacturer Code:NBP177044
Catalog # NBP177044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TACE. The immunogen is located within the last 50 amino acids of TACE. Amino Acid Squence: ASFKLQRQNRVDSKETE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAM 17; ADAM 17 ADAM metallopeptidase domain 17 ADAM metallopeptidase domain 18 ADAM18 CD156B CD156b antigen CSVP disintegrin and metalloproteinase domain-containing protein 17 EC 3.4.24.86 MGC71942 Snake venom-like protease TACEcSVP TNF-alpha convertase TNF-alpha converting enzyme TNF-alpha-converting enzyme tumor necrosis factor alpha converting enzyme; ADAM metallopeptidase domain 18; CD156b; Disintegrin and metalloproteinase domain-containing protein 17; Snake venom-like protease; TNF-alpha convertase; TNF-alpha converting enzyme; TNF-alpha-converting enzyme; tumor necrosis factor, alpha, converting enzyme
Gene Aliases: ADAM17; ADAM18; CD156B; CSVP; NISBD; NISBD1; TACE
UniProt ID: (Human) P78536
Entrez Gene ID: (Human) 6868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.