R&D Systems
Manufacturer Code:AF2129SP
Catalog # AF2129SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human TACE Asp695-Cys824 Accession # P78536 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAM 17; ADAM 17 ADAM metallopeptidase domain 17 ADAM metallopeptidase domain 18 ADAM17 ADAM18 CD156b CD156b antigen CSVP disintegrin and metalloproteinase domain-containing protein 17 EC 3.4.24.86 MGC71942 Snake venom-like protease TACEcSVP TNF-alpha convertase TNF-alpha converting enzyme TNF-alpha-converting enzyme tumor necrosis factor alpha converting enzyme; ADAM metallopeptidase domain 18; CD156b; Disintegrin and metalloproteinase domain-containing protein 17; Snake venom-like protease; TNF-alpha convertase; TNF-alpha converting enzyme; TNF-alpha-converting enzyme; tumor necrosis factor, alpha, converting enzyme
Gene Aliases: ADAM17; ADAM18; CD156B; CSVP; NISBD; NISBD1; TACE
UniProt ID: (Human) P78536
Entrez Gene ID: (Human) 6868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.