Novus Biologicals
Manufacturer Code:NBP267671
Catalog # NBP267671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SN73-05 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ERIC-1; ERIC1ERIC-1 MGC117382 MGC133242 transforming acidic coiled-coil-containing protein 3 transforming acidic coiled-coil containing protein 3; Transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3
Gene Aliases: ERIC-1; ERIC1; TACC3
UniProt ID: (Human) Q9Y6A5
Entrez Gene ID: (Human) 10460
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.