OriGene
Manufacturer Code:TA501264
Catalog # 50-167-2407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6H4 |
Immunogen | Full length human recombit protein of human TACC3 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ERIC-1; ERIC-1 ERIC1; Transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3
Gene Aliases: ERIC-1; ERIC1; TACC3
UniProt ID: (Human) Q9Y6A5
Entrez Gene ID: (Human) 10460
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.