OriGene
Manufacturer Code:TA804400
Catalog # 50-168-0230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8A5 |
Immunogen | Human recombit protein fragment corresponding to amino acids 727-1026 of human TACC2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: anti zuai-1; Anti-Zuai-1; AZU-1; AZU-1 ECTACC; Transforming acidic coiled-coil-containing protein 2; transforming, acidic coiled-coil containing protein 2
Gene Aliases: AZU-1; ECTACC; TACC2
UniProt ID: (Human) O95359
Entrez Gene ID: (Human) 10579
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.