Abnova
Manufacturer Code:PAB24942
Catalog # 89-126-618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 1-11 of rat Tac1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Guinea pig polyclonal antibody raised against synthetic peptide of Tac1.
For Research Use Only
Protein Aliases: C-terminal-flanking peptide; Neurokinin A; Neuromedin L; Neuropeptide gamma; Neuropeptide K; NKA; NPK; PPT; Ppt5fl; PPTA3; Protachykinin-1; RATPPTA3; Substance K; Substance P; TAC; Tachykinin (substance P, neurokinin A, neuropeptide K, neuropeptide gamma); tachykinin 1
Gene Aliases: Nka; Nkna; Ppt5fl; PPTA3; RATPPTA3; TAC; Tac1; Tac2
UniProt ID: (Rat) P06767
Entrez Gene ID: (Rat) 24806
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.