Invitrogen
Manufacturer Code:PA5114782
Catalog # PIPA5114782
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAB3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4921526G09Rik Kiaa4135 MAP3K7IP3 mitogen activated protein kinase kinase kinase 7 interacting protein 3 mitogen-activated protein kinase kinase kinase 7 interacting protein 3 Mitogen-activated protein kinase kinase kinase 7-interacting protein 3 mKIAA4135 NAP1 NF-kappa-B-activating protein 1 NFkB activating protein 1 Tab3 TAB-3 TAK1-binding protein 3 TGF-beta activated kinase 1/MAP3K7 binding protein 3 TGF-beta-activated kinase 1 and MAP3K7-binding protein 3 TGF-beta-activated kinase 1-binding protein 3; mitogen activated protein kinase kinase kinase 7 interacting protein 3; mitogen-activated protein kinase kinase kinase 7 interacting protein 3; Mitogen-activated protein kinase kinase kinase 7-interacting protein 3; NF-kappa-B-activating protein 1; NFkB activating protein 1; TAB-3; TAK1-binding protein 3; TGF-beta-activated kinase 1 and MAP3K7-binding protein 3; TGF-beta-activated kinase 1-binding protein 3
Gene Aliases: 4921526G09Rik; Kiaa4135; MAP3K7IP3; mKIAA4135; NAP1; TAB3
UniProt ID: (Human) Q8N5C8, (Mouse) Q571K4
Entrez Gene ID: (Human) 257397, (Mouse) 66724
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.