Invitrogen
Manufacturer Code:PA528683
Catalog # PIPA528683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 210 and 504 of Human TAB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3&prime-Tab1 MAP3K7IP1 TAK1-binding protein 1 TGF-beta-activated kinase 1 and MAP3K7-binding protein 1 mitogen-activated protein kinase kinase kinase 7-interacting protein 1 transforming growth factor beta-activated kinase-binding protein 1; beta activated kinase-1 binding protein-1; mitogen activated protein kinase kinase kinase 7 interacting protein 1; mitogen-activated protein kinase kinase kinase 7 interacting protein 1; Mitogen-activated protein kinase kinase kinase 7-interacting protein 1; TAK1-binding protein 1; TGF-beta-activated kinase 1 and MAP3K7-binding protein 1; TGF-beta-activated kinase 1-binding protein 1; transforming growth factor beta-activated kinase-binding protein 1
Gene Aliases: 2310012M03Rik; 3'-Tab1; b2b449Clo; MAP3K7IP1; TAB1
UniProt ID: (Human) Q15750, (Mouse) Q8CF89
Entrez Gene ID: (Human) 10454, (Mouse) 66513
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.