Invitrogen
Manufacturer Code:PA5116001
Catalog # PIPA5116001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAB1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310012M03Rik 3'-Tab1 b2b449Clo beta activated kinase-1 binding protein-1 MAP3K7IP1 MGC57664 mitogen activated protein kinase kinase kinase 7 interacting protein 1 mitogen-activated protein kinase kinase kinase 7 interacting protein 1 Mitogen-activated protein kinase kinase kinase 7-interacting protein 1 RP3-407F17.2 Tab1 TAB1beta Tak1-binding protein 1 TGF-beta activated kinase 1/MAP3K7 binding protein 1 TGF-beta-activated kinase 1 and MAP3K7-binding protein 1 TGF-beta-activated kinase 1-binding protein 1 transforming growth factor beta-activated kinase-binding protein 1; beta activated kinase-1 binding protein-1; mitogen activated protein kinase kinase kinase 7 interacting protein 1; mitogen-activated protein kinase kinase kinase 7 interacting protein 1; Mitogen-activated protein kinase kinase kinase 7-interacting protein 1; TAK1-binding protein 1; TGF-beta-activated kinase 1 and MAP3K7-binding protein 1; TGF-beta-activated kinase 1-binding protein 1; transforming growth factor beta-activated kinase-binding protein 1
Gene Aliases: 2310012M03Rik; 3'-Tab1; b2b449Clo; MAP3K7IP1; TAB1
UniProt ID: (Human) Q15750, (Mouse) Q8CF89
Entrez Gene ID: (Human) 10454, (Mouse) 66513, (Rat) 315139
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.