Novus Biologicals
Manufacturer Code:NBP121405
Catalog # NBP121405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 435 and 485 of human TAK1-binding protein 1 using the numbering given in entry NP_006107.1 (GeneID 10454). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP3K7IP13'-Tab1 MGC57664 mitogen-activated protein kinase kinase kinase 7 interacting protein 1 Mitogen-activated protein kinase kinase kinase 7-interacting protein 1 TAK1-binding protein 1 TGF-beta activated kinase 1/MAP3K7 binding protein 1 TGF-beta-activated kinase 1 and MAP3K7-binding protein 1 TGF-beta-activated kinase 1-binding protein 1 transforming growth factor beta-activated kinase-binding protein 1; Mitogen-activated protein kinase kinase kinase 7-interacting protein 1; TAK1-binding protein 1; TGF-beta-activated kinase 1 and MAP3K7-binding protein 1; TGF-beta-activated kinase 1-binding protein 1; transforming growth factor beta-activated kinase-binding protein 1
Gene Aliases: 3'-Tab1; MAP3K7IP1; TAB1
UniProt ID: (Human) Q15750
Entrez Gene ID: (Human) 10454
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.