R&D Systems
Manufacturer Code:AF3578
Catalog # AF3578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human TAB1 Ser7-Glu159 Accession # Q15750 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MAP3K7IP1 MAP3K7IP13'-Tab1 MGC57664 mitogen-activated protein kinase kinase kinase 7 interacting protein 1 Mitogen-activated protein kinase kinase kinase 7-interacting protein 1 TAK1-binding protein 1 TGF-beta activated kinase 1/MAP3K7 binding protein 1 TGF-beta-activated kinase 1 and MAP3K7-binding protein 1 TGF-beta-activated kinase 1-binding protein 1 transforming growth factor beta-activated kinase-binding protein 1; Mitogen-activated protein kinase kinase kinase 7-interacting protein 1; TAK1-binding protein 1; TGF-beta-activated kinase 1 and MAP3K7-binding protein 1; TGF-beta-activated kinase 1-binding protein 1; transforming growth factor beta-activated kinase-binding protein 1
Gene Aliases: 3'-Tab1; MAP3K7IP1; TAB1
UniProt ID: (Human) Q15750
Entrez Gene ID: (Human) 10454
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.