Abnova
Manufacturer Code:PAB27703
Catalog # 89-129-220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 16 amino acid at cytoplasmic domain of human TAAR8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TAAR8.
For Research Use Only
Protein Aliases: G protein-coupled receptor 102; G-protein coupled receptor 102; GPR102; TA5; TaR-5; TaR-8; TAR5; Trace amine receptor 5; trace amine receptor 8; Trace amine-associated receptor 8; TRAR5
Gene Aliases: GPR102; TA5; TAAR8; TaR-5; TaR-8; TAR5; TRAR5
UniProt ID: (Human) Q969N4
Entrez Gene ID: (Human) 83551
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.