Abnova
Manufacturer Code:H00083551K
Catalog # 89-204-370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human TAAR8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human TAAR8 peptide using ARM Technology.
For Research Use Only
Protein Aliases: G protein-coupled receptor 102; G-protein coupled receptor 102; GPR102 TA5 TAR5 TRAR5; TaR-5; TaR-8; Trace amine receptor 5; trace amine receptor 8; Trace amine-associated receptor 8
Gene Aliases: GPR102; TA5; TAAR8; TaR-5; TaR-8; TAR5; TRAR5
UniProt ID: (Human) Q969N4
Entrez Gene ID: (Human) 83551
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.