Invitrogen
Manufacturer Code:OSR00116W
Catalog # PIOSR00116W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from mouse TAAR7a b e f conjugated to an immunugenic carrier protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mTaar7a; TaR-7a; TaR-8; trace amine receptor 7a; Trace amine receptor 8; Trace amine-associated receptor 7a; Trace amine-associated receptor 7a Taar7a Trace amine-associated receptor 7b Taar7b Trace amine-associated receptor 7e Taar7e Trace amine-associ; trace-amine-associated receptor 7a
Gene Aliases: Gm229; Ta8; Taar7a; taR-7a; Tar8; Trar8
UniProt ID: (Mouse) Q5QD12, (Rat) Q923Y2
Entrez Gene ID: (Mouse) 215856, (Rat) 294125
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.