Invitrogen
Manufacturer Code:PA570687
Catalog # PIPA570687
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human TAAR6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mTaar6; TAAR6 TA4 TaR-4 TAR4 TAR6 TaR-6 TRAR4 Trace amine receptor 4 Trace amine receptor 6 RP11-295F4.3; TaR-4; TaR-6; Trace amine receptor 4; trace amine receptor 6; Trace amine-associated receptor 6; trace-amine-associated receptor 6
Gene Aliases: Gm228; TA4; TAAR6; taR-4; taR-6; TAR4; TAR6; TRAR4
UniProt ID: (Human) Q96RI8, (Rat) Q923Y5, (Mouse) Q5QD13
Entrez Gene ID: (Human) 319100, (Bovine) 783342, (Rabbit) 100343908, (Rat) 294124, (Mouse) 215855
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.