Invitrogen
Manufacturer Code:PA545005
Catalog # PIPA545005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human TAAR5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTaar5; mTaar5; Putative neurotransmitter receptor; TaR-5; Trace amine associated receptor 5 MGC138414 MGC138416 PNR RP11-295F4.5; trace amine receptor 5; Trace amine-associated receptor 5; Trimethylamine receptor
Gene Aliases: Gm227; PNR; TAAR5; taR-5
UniProt ID: (Human) O14804, (Rat) Q5QD23, (Mouse) Q5QD14
Entrez Gene ID: (Horse) 100073122, (Human) 9038, (Rabbit) 100344162, (Rat) 294123, (Mouse) 215854
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.