Invitrogen
Manufacturer Code:PA533067
Catalog # PIPA533067
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from C-Terminus of human TAAR5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hTaar5; PNR RP11-295F4.5 putative neurotransmitter receptor taR-5 trace amine receptor 5 trace amine-associated receptor 5; Putative neurotransmitter receptor; TaR-5; trace amine receptor 5; Trace amine-associated receptor 5
Gene Aliases: PNR; TAAR5
UniProt ID: (Human) O14804
Entrez Gene ID: (Human) 9038
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.