Novus Biologicals
Manufacturer Code:NBP286840
Catalog # NB034462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human TAAR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 58; G protein-coupled receptor 58 GPR58taR-2 G-protein coupled receptor 58 TaR-2 trace amine associated receptor 2 Trace amine receptor 2 trace amine-associated receptor 2; G-protein coupled receptor 58; TaR-2; Trace amine-associated receptor 2
Gene Aliases: GPR58; TAAR2; taR-2
UniProt ID: (Human) Q9P1P5
Entrez Gene ID: (Human) 9287
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.