Invitrogen
Manufacturer Code:PA533066
Catalog # PIPA533066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from 3rd cytoplasmic domain of human TAAR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RP11-295F4.9 TA1 TAR1 TRAR1 taR-1 trace amine receptor 1 trace amine-associated receptor 1; TaR-1; trace amine receptor 1; Trace amine-associated receptor 1
Gene Aliases: TA1; TAAR1; TAR1; TRAR1
UniProt ID: (Human) Q96RJ0
Entrez Gene ID: (Human) 134864
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.