Invitrogen
Manufacturer Code:PA5115999
Catalog # PIPA5115999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TAAR1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC126874 MGC138399 RP11-295F4.9 RTAR1 Snorf33 Ta1 TAAR1 TAR1 TaR-1 trace amine associated receptor 1 trace amine receptor 1 trace amine-associated receptor 1 trace-amine-associated receptor 1 Trar1; TaR-1; trace amine receptor 1; Trace amine-associated receptor 1
Gene Aliases: TA1; TAAR1; taR-1; TAR1; TRAR1
UniProt ID: (Human) Q96RJ0, (Rat) Q923Y9, (Mouse) Q923Y8
Entrez Gene ID: (Human) 134864, (Rat) 113914, (Mouse) 111174
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.