Invitrogen
Manufacturer Code:OSR00079W
Catalog # PIOSR00079W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from human TAAR1 conjugated to an immunogenic carrier Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TaR-1; trace amine receptor 1; Trace amine receptor 1 TaR-1 TA1 TAR1 TRAR1; Trace amine-associated receptor 1
Gene Aliases: TA1; TAAR1; TAR1; TRAR1
UniProt ID: (Human) Q96RJ0
Entrez Gene ID: (Human) 134864
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.