Abnova
Manufacturer Code:PAB0313
Catalog # 89-114-132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids 200-230 of human TAAR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant TAAR1.
For Research Use Only
Protein Aliases: MGC126874; MGC138399; RP11-295F4.9; TA1; TaR-1; TAR1; trace amine receptor 1; Trace amine-associated receptor 1; TRAR1
Gene Aliases: TA1; TAAR1; TAR1; TRAR1
UniProt ID: (Human) Q96RJ0
Entrez Gene ID: (Human) 134864
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.