Invitrogen
Manufacturer Code:12582580
Catalog # 50-108-93
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | eBio4B10 (4B10) |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor TBX21; T-cell-specific T-box transcription factor T-bet; T-PET T-bet TBET TBLYM T-box expressed in T cells T-box protein 21 T-box transcription factor TBX21 T-cell-specific T-box transcription factor T-bet transcription factor TBLYM; Transcription factor TBLYM
Gene Aliases: T-bet; T-PET; TBET; TBLYM; TBT1; TBX21
UniProt ID: (Human) Q9UL17, (Mouse) Q9JKD8
Entrez Gene ID: (Rhesus monkey) 694044, (Human) 30009, (Mouse) 57765
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.