Abnova
Manufacturer Code:MAB18227
Catalog # 89-338-152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S275-14 |
Immunogen | Recombinant protein corresponding to amino acids 150-239 at cytoplasmic C2A domain of mouse Syt7. |
Conjugate | ATTO633 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI851541; B230112P13Rik; synaptotagmin 7; Synaptotagmin VII; Synaptotagmin-7; SytVII
Gene Aliases: AI851541; B230112P13Rik; Syt7
UniProt ID: (Mouse) Q9R0N7
Entrez Gene ID: (Mouse) 54525
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.