Novus Biologicals
Manufacturer Code:NB1001346
Catalog # NB1001346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence ASGRRAPRTGLLE-C corresponding to N-Terminus according to NP_003089.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 59 kDa dystrophin-associated protein A1 acidic component 1; 59 kDa dystrophin-associated protein A1 acidic component 1 dJ1187J4.5 LQT12 Pro-TGF-alpha cytoplasmic domain-interacting protein 1 SNT1alpha-1-syntrophin syntrophin alpha 1 (dystrophin-associated protein A1 59kD acidic component) syntrophin alpha 1 (dystrophin-associated protein A1 59kDa acidic component) syntrophin-1 TACIP1acidic alpha 1 syntrophin; acidic alpha 1 syntrophin; Alpha-1-syntrophin; dystrophin-associated protein A1, 59kDa, acidic component; Pro-TGF-alpha cytoplasmic domain-interacting protein 1; syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component); Syntrophin-1; TACIP1
Gene Aliases: dJ1187J4.5; LQT12; SNT1; SNTA1; TACIP1
UniProt ID: (Human) Q13424
Entrez Gene ID: (Human) 6640
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.