Invitrogen
Manufacturer Code:PA528826
Catalog # PIPA528826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 234 and 298 of Human Syntenin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MDA-9; MDA-9 ST1 SYCL TACIP18 melanoma differentiation associated protein-9 melanoma differentiation-associated protein 9 pro-TGF-alpha cytoplasmic domain-interacting protein 18 scaffold protein Pbp1 syndecan-binding protein 1 syntenin-1; melanoma differentiation associated protein-9; Melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; Scaffold protein Pbp1; syndecan binding protein (syntenin); syndecan interacting protein; Syndecan-binding protein 1; syntenin; Syntenin-1; TACIP18
Gene Aliases: MDA-9; MDA9; SDCBP; ST1; SYCL; syntenin-1; TACIP18
UniProt ID: (Human) O00560, (Mouse) O08992, (Rat) Q9JI92
Entrez Gene ID: (Human) 6386, (Mouse) 53378, (Rat) 83841
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.