Novus Biologicals
Manufacturer Code:NBP192469
Catalog # NBP192469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GMEMDYEEVIRLLEAKITELKAQLADYSDQNKESVQDLKKRIMVLDCQLRKSEMARKTFEASTEKLLHFVEAIQEVFSDNSTPLSNLSERRAVLASQT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ16496 MGC50337 STX4-interacting protein SynipMGC149829 syntaxin 4 interacting protein Syntaxin 4-interacting protein syntaxin binding protein 4 syntaxin-binding protein 4; STX4-interacting protein; syntaxin 4 interacting protein; Syntaxin 4-interacting protein; Syntaxin-binding protein 4
Gene Aliases: STXBP4; Synip
UniProt ID: (Human) Q6ZWJ1
Entrez Gene ID: (Human) 252983
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.